RNA isolation, total RNA kit, E-Z 96®
Supplier: OMEGA BIO-TEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R1034-02 R1034-00
R1034-02EA 2660 GBP
R1034-02 R1034-00
RNA isolation, total RNA kit, E-Z 96®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E-Z 96® Total RNA Kit is designed for isolation of total cellular RNA from up to 5×10⁶ cultured cells or soft tissues. This kit can process single or multiple samples in less than 60 minutes. Utilising HiBind® silica plate technology, the need for phenol/chloroform extractions, CsCl gradient ultracentrifugation, and precipitation with isopropanol or LiCl are eliminated. Samples are lysed in a denaturing lysis buffer which inactivates RNases. Binding conditions are adjusted and the lysate is transferred to a 96-well HiBind® RNA Plate where the RNA is purified via three wash steps. High quality RNA is eluted in RNase-free water. RNA purified using the E-Z 96® Total RNA method is ready for applications such as RT-PCR, qPCR, differential display, microarrays, and other downstream applications.