dNTP Mix, 10 mM

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

R0191 R0192 R0193
R0191EA 34.9 GBP
R0191 R0192 R0193
dNTP Mix, 10 mM
Nucleic Acid Reagents Nucleotides

dNTP mix contains premixed aqueous solutions of dATP, dCTP, dGTP and dTTP, each at a final concentration of 10 mM.


  • Highly stable
  • Stable for years at –20 °C
  • Stable after multiple freeze-thaw cycles
  • Up to 95% of dNTPs remain in triphosphate form even after 7 weeks at room temperature
  • Up to 90% of dNTPs remain in triphosphate form after 30 cycles of PCR (1 minute at 94 °C, 3 minutes at 72 °C)


The nucleotides have greater than 99% purity, are free of nuclease activities, human and E. coli DNA. Mixes offer the possibility to reduce the number of pipetting steps and the risk of reaction set up errors. They are designed for many different molecular biology applications.


For use in all molecular biology applications, including PCR, real-time PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, RDA, MDA, DNA labeling, and DNA sequencing.


Caution: For research use only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR