Variant of concern detection kits, SNPsig®
Supplier: PRIMERDESIGN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R00130 R00131 R00135 R00134 R00132 R00137 R00146 R00147 R00149 R00152
R00130EA 0 GBP
R00130 R00131 R00135 R00134 R00132 R00137 R00146 R00147 PRDER00149 PRDER00152
Variant of concern detection kits, SNPsig®
Clinical Diagnostic Supplies Clinical Reagents and Kits
These high quality assays detect the SARS-CoV-2 variants of concern to give fast and clear results to customers.
  • Rapid detection of specific detection profiles
  • High priming efficiency
  • Sensitive to <100 copies of target
  • No cold chain shipping
  • Open PCR platform compatibility
  • Lyophilised components
An alternative to next generation sequencing and S gene target failure (SGTF), it enables scientists to analyse and monitor these specific genomic mutations. With a streamlined workflow for easy use with readily prepared mastermix, these kits are pivotal in the role of screening for SARS-CoV-2 variants for the purpose of genomic surveillance studies.
Accessories information: Accessories not included.
Delivery information: Shipped at ambient temperatures due to lyophilsed kit contents. Complete kit. Supplied with ready made mastermix, positive control templates for both mutant and wild-type, preparation buffers and genotyping primer/probe mix.
Caution: Research use only kits.