Standard ground joints
Supplier: QUARTZTECHNISCHE WERKSTATTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1044 1047 1030 1045 1031 1033 1032 1046 1043
QUTW1044EA 98.7 GBP
QUTW1044 QUTW1047 QUTW1030 QUTW1045 QUTW1031 QUTW1033 QUTW1032 QUTW1046 QUTW1043
Standard ground joints
Adapters Adapters, Conical Ground Joints
Connections, quartz glass, with standard ground joint according to DIN 12 249.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving high resistance to temperature changes.