Dissolution basket shafts, Apparatus 1, QLA
Supplier: QUALITY LAB ACCESSORIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BSDRNG-16 BSKSHT-13 BSKSHT-15 BSKSHT-16 BSKSHT-19 PADHLD-12A PADHLD-16A TUMSCR-DK
QLAABSDRNG-16EA 319 GBP
QLAABSDRNG-16 QLAABSKSHT-13 QLAABSKSHT-15 QLAABSKSHT-16 QLAABSKSHT-19 QLAAPADHLD-12A QLAAPADHLD-16A QLAATUMSCR-DK
Dissolution basket shafts, Apparatus 1, QLA
Dissolution Apparatus
These Clip style and O-ring Push-on style basket shafts are ideal for most dissolution baths.
  • Made of high quality 316 electropolished stainless steel
  • Laser marked with a unique serial number
  • Spring clips on clip style basket shafts
Clip style shafts utilise a proprietary, spring tempered steel clip, which retains the original gripping tension far longer than conventional shafts and comply with USP (711) and Original Equipment Manufacturer's dimensional specifications. O-ring push-on style shafts comply with Original Equipment Manufacturer'sMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable