Anti-Mycoplasma bovis Mouse Monoclonal Antibody

Supplier: QED Bioscience
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

18201
QEDB18201EA 380 GBP
QEDB18201
Anti-Mycoplasma bovis Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Mycoplasma bovis
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR