Safety spectacles, PW30
Supplier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PW30CLR
PRTWPW30CLREA 7.8 GBP
PRTWPW30CLR
Safety spectacles, PW30
Safety Glasses
Metal free safety glasses made from impact resistant PC offering excellent protection and comfort.
  • Moulded in brow guard
  • Vented side shields
Certifications: EN 166 1F AS