Disposable particle filtering respirators, FFP2, P201, BizTex®
Supplier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
P201WHR
PRTWP201WHREA 48.6 GBP
PRTWP201WHR
Disposable particle filtering respirators, FFP2, P201, BizTex®
Respirators Disposable Respirators Particulate Respirators
BizTex consists of a polypropylene substrate to which a laminated microporous PE film is applied (PP/PE). The outer layer of fabric prevents the passage of liquid molecules inwards yet permits the vapour molecules that build up inside to pass through. Mask protects against fine toxic dust, fumes and water based mists.
  • Adjustable nose bridge
  • Sweat absorbing nose foam (silicone/latex-free)
  • Twin elastic headband
  • Colour coded to indicate performance level
  • With easy flow exhalation valve
  • Can be used in concentrations of contaminants up to 10 times WEL
Certifications: EN 149:2001+A1:2009
Order Now
SPECIFICATIONS
Environmentally Preferable