Thulium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PTMN-100 VHG-PTMN-500 VHG-TTMN-100 VHG-TTMN-500
PROOVHG-PTMN-100EA 125 GBP
PROOVHG-PTMN-100 PROOVHG-PTMN-500 PROOVHG-TTMN-100 PROOVHG-TTMN-500
Thulium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standards Cationic Standards Thulium Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.