Neodymium standard solution, for ICP, in 5% HNO₃, VHG
Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PNDN-100 VHG-PNDN-500 VHG-TNDN-100 VHG-TNDN-500
PROOVHG-PNDN-100EA 90.8 GBP
PROOVHG-PNDN-100 PROOVHG-PNDN-500 PROOVHG-TNDN-100 PROOVHG-TNDN-500
Neodymium standard solution, for ICP, in 5% HNO₃, VHG
Standards Cationic Standards Neodymium Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.