Molybdenum standard solution, for ICP, in 5% HNO₃, VHG
Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PMONF-100 VHG-PMONF-50 VHG-PMONF-500 VHG-TMONF-100 VHG-TMONF-500
PROOVHG-PMONF-100EA 76.9 GBP
PROOVHG-PMONF-100 PROOVHG-PMONF-50 PROOVHG-PMONF-500 PROOVHG-TMONF-100 PROOVHG-TMONF-500
Molybdenum standard solution, for ICP, in 5% HNO₃, VHG
Standards Cationic Standards Molybdenum Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.