Boron standard solution, for ICP, in NH₄OH, VHG
Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PBZ-100 VHG-PBZ-500 VHG-TBZ-100 VHG-TBZ-500
PROOVHG-PBZ-100EA 80 GBP
PROOVHG-PBZ-100 PROOVHG-PBZ-500 PROOVHG-TBZ-100 PROOVHG-TBZ-500
Boron standard solution, for ICP, in NH₄OH, VHG
Standards Cationic Standards Boron Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.