Tin standard solution, for ICP, in 5% HNO₃, trace HF, CRM, VHG
Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PSNNF-100 VHG-LSNNF-100 VHG-PSNNF-50 VHG-PSNNF-500 VHG-TSNNF-100 VHG-TSNNF-500
PROOVHG-PSNNF-100EA 76.9 GBP
PROOVHG-PSNNF-100 PROOVHG-LSNNF-100 PROOVHG-PSNNF-50 PROOVHG-PSNNF-500 PROOVHG-TSNNF-100 PROOVHG-TSNNF-500
Tin standard solution, for ICP, in 5% HNO₃, trace HF, CRM, VHG
Standards Cationic Standards Tin Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.