Platinum standard solution, for ICP, in 20% HCl, VHG
Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PPTH-100 VHG-PPTH-50 VHG-PPTH-500 VHG-TPTH-100 VHG-TPTH-500
PROOVHG-PPTH-100EA 117 GBP
PROOVHG-PPTH-100 PROOVHG-PPTH-50 PROOVHG-PPTH-500 PROOVHG-TPTH-100 PROOVHG-TPTH-500
Platinum standard solution, for ICP, in 20% HCl, VHG
Standards Cationic Standards Platinum Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.