Petroleum standards, heavy mineral oils, CRM
Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-OIL-75-500
PROOVHG-OIL-75-500EA 75.4 GBP
PROOVHG-OIL-75-500
Petroleum standards, heavy mineral oils, CRM
Standards Petroleum Industry Standards
This matrix material solution is intended for use as a diluent for other metallo-organic standards.
Certifications: ISO 9001 and ISO/IEC 17025 Certified and traceable to NIST.