Zinc standard solution, for ICP, in HNO₃, CRM, VHG
Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PZNN-100 VHG-PZNN-500 VHG-LZNN-100 VHG-LZNN-500 VHG-PZNN-250 VHG-PZNN-50 VHG-TZNN-100 VHG-TZNN-500
PROOVHG-PZNN-100EA 76.9 GBP
PROOVHG-PZNN-100 PROOVHG-PZNN-500 PROOVHG-LZNN-100 PROOVHG-LZNN-500 PROOVHG-PZNN-250 PROOVHG-PZNN-50 PROOVHG-TZNN-100 PROOVHG-TZNN-500
Zinc standard solution, for ICP, in HNO₃, CRM, VHG
Standards Cationic Standards Zinc Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.