Nitrate(N), ion chromatography standard, VHG
Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-INO3N-500 VHG-INO3N-100
PROOVHG-INO3N-500EA 170 GBP
PROOVHG-INO3N-500 PROOVHG-INO3N-100
Nitrate(N), ion chromatography standard, VHG
Standards Anionic Standards Nitrate(N) Standards
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certifications: Traceable to NIST and SRM.