Zinc oxide

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

NCSGC76207
PROONCSGC76207EA 586 GBP
PROONCSGC76207
Zinc oxide
Zinc oxide
Formula: ZnO
MW: 81.39 g/mol
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR