Dibutyl sebacate

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

MM0925.00
PROOMM0925.00EA 2110 GBP
PROOMM0925.00
Dibutyl sebacate
Dibutyl sebacate
Formula: C₁₈H₃₄O₄
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: –11 °C
Density: 0,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00027218
CAS Number: 109-43-3
EINECS: 203-672-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR