Chloramphenicol

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

MM0516.00
PROOMM0516.00EA 1850 GBP
PROOMM0516.00
Chloramphenicol
Chloramphenicol
Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 150 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR