4-Chloro-3-sulphamoylbenzoic acid

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MM0477.04
PROOMM0477.04EA 766 GBP
PROOMM0477.04
4-Chloro-3-sulphamoylbenzoic acid
4-Chloro-3-sulphamoylbenzoic acid
Formula: C₇H₆ClNO₄S
MDL Number: MFCD00012375
CAS Number: 1205-30-7
EINECS: 214-882-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR