Acetophenone

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Methyl phenyl ketone, Phenyl methyl ketone

MM0237.01
PROOMM0237.01EA 914 GBP
PROOMM0237.01
Acetophenone
Acetophenone
Formula: C₈H₈O
Boiling Pt: 201.7 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.027 g/cm³ (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00008724
CAS Number: 98-86-2
EINECS: 202-708-7
UN: 3334
ADR: 9,
Merck Index: 13,00074

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR