2-Aminobenzyl alcohol

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (2-Aminophenyl)methanol, 2-Hydroxymethylaniline, 2-(Hydroxymethyl)aniline, 2-Aminobenzyl Alcohol

MM0147.04
PROOMM0147.04EA 1120 GBP
PROOMM0147.04
2-Aminobenzyl alcohol
2-Aminobenzyl alcohol
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 275
Melting Pt: 81
Storage Temperature: Refrigerator
MDL Number: MFCD00007749
CAS Number: 5344-90-1
EINECS: 226-293-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR