Carbazole

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

ERM-FC021
PROOERM-FC021EA 430 GBP
PROOERM-FC021
Carbazole
Carbazole
Formula: C₁₂H₉N
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 236 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00004960
CAS Number: 86-74-8
EINECS: 201-696-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR