1,2-Dichloroethylene-[D₂] (98% D)

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1,2-Dichloroethylene-[²H₂]

CILDLM19360.1 CIL-DLM-1936-1
PROOCILDLM19360.1EA 353 GBP
PROOCILDLM19360.1 PROOCIL-DLM-1936-1
1,2-Dichloroethylene-[D₂] (98% D)
1,2-Dichloroethylene-[D2]
Formula: C₂D₂Cl₂
CAS Number: 15075-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR