Acridine-[D₉] (98% D)

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Acridine-[²H₉]

CILDLM-849-0.1 CILDLM-849-0.5
PROOCILDLM-849-0.1EA 793 GBP
PROOCILDLM-849-0.1 PROOCILDLM-849-0.5
Acridine-[D₉] (98% D)
Acridine-[D9]
Formula: C₁₃D₉N
CAS Number: 34749-75-2
UN: 2713
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR