2,2',3,4,4',5,6-Heptachlorobiphenyl (PCB No. 181)

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CIL-PCB-181-C
PROOCIL-PCB-181-CEA 1090 GBP
PROOCIL-PCB-181-C
2,2',3,4,4',5,6-Heptachlorobiphenyl (PCB No. 181)
2,2',3,4,4',5,6-Heptachlorobiphenyl (PCB No. 181)
Formula: (C₆H₃Cl₂)C₆Cl₅
MW: 395.32 g/mol
CAS Number: 74472-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR