n-Octadecane

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Octadecane

CHE170
PROOCHE170EA 314 GBP
PROOCHE170
n-Octadecane
n-Octadecane
Formula: H₃C(CH₂)₁₆CH₃
MW: 254.5 g/mol
Boiling Pt: 316.1
Melting Pt: 26…29 °C
Density: 0.7765 g/cm³ (20 °C)
Flash Pt: 165
Storage Temperature: Ambient
MDL Number: MFCD00009007
CAS Number: 593-45-3
EINECS: 209-790-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR