n-Tridecane

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Tridecane

CHE153
PROOCHE153EA 314 GBP
PROOCHE153
n-Tridecane
n-Tridecane
Formula: H₃C(CH₂)₁₁CH₃
MW: 184.37 g/mol
Boiling Pt: 235.4
Melting Pt: –5.5 °C
Density: 0.7564 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Ambient
MDL Number: MFCD00008979
CAS Number: 629-50-5
EINECS: 211-093-4
UN: 3295
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR