Propylbenzene

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

CHE121
PROOCHE121EA 314 GBP
PROOCHE121
Propylbenzene
Propylbenzene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 159
Melting Pt: –99 °C
Density: 0.862 g/cm³ (20 °C)
Flash Pt: 45
Storage Temperature: Ambient
MDL Number: MFCD00009377
CAS Number: 103-65-1
EINECS: 203-132-9
UN: 2364
ADR: 3 III
Merck Index: 12,08029

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR