Dexamethasone

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion

BP578
PROOBP578EA 459 GBP
PROOBP578
Dexamethasone
Dexamethasone
Formula: C₂₂H₂₉FO₅
MW: 392.47 g/mol
Melting Pt: 268…271 °C
MDL Number: MFCD00064136
CAS Number: 50-02-2
EINECS: 200-003-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR