Bulb pipette, FORTUNA®, class AS
Supplier: Poulten & Graf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.2323104 1.2325102 1.2323404 1.2323504 1.2324502
POUL1.2323104EA 0 GBP
POUL1.2323104 POUL1.2325102 POUL1.2323404 POUL1.2323504 POUL1.2324502
Bulb pipette, FORTUNA®, class AS
Pipettes Bulb Pipettes
Clear glass.
  • Two marks
  • Blue or amber graduation
  • Conformity approved, with batch certificate