Plum iBox
Supplier: Plum Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4921 4922 4920 4931 4923 4900
PLUM4921EA 83.7 GBP
PLUM4921 PLUM4922 PLUM4920 PLUM4931 PLUM4923 PLUM4900
Plum iBox
First Aid Supplies Eyewash Stations and Eyewash Bottles
The new Plum iBox is balancing key features in a modern design and is built in a strong and durable material. See-through window lets users keep track of expiry dates immediately.
  • Clearly visible eyewash bottles
  • Quick and efficient overview of expiry dates and product condition
  • Grab from the wall bracket and quickly bring to injured person
  • Spacious design allows access to bottles fast and easily
  • Easily opens – mounted on the wall or standing on the floor
  • Strong and durable material (EPP)
Release Plum iBox from the wall and quickly bring the eyewash toMore Product Information