Sodium deoxycholate

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

89904 89905
PIER89904EA 179 GBP
PIER89904 PIER89905
Sodium deoxycholate
Sodium deoxycholate
Formula: C₂₄H₃₉NaO₄
Melting Pt: 357…365 °C
Storage Temperature: Ambient
MDL Number: MFCD00064139
CAS Number: 302-95-4
EINECS: 206-132-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR