Cell-free protein expression vectors, T7, mammalian
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
88860 88865 88866 88868 88869 88870 88871
PIER88860EA 136 GBP
PIER88860 PIER88865 PIER88866 PIER88868 PIER88869 PIER88870 PIER88871
Cell-free protein expression vectors, T7, mammalian
Vectors, Plasmids and Libraries
The T7 cell-free expression vectors (pT7CFE1) are cloning plasmids optimised for use with the Thermo Scientific™ 1-Step Human In Vitro Protein Expression System. They are used for the in vitro translation (IVT) of genes into functional proteins when used in combination with the 1-Step Human IVT system. The pT7CFE1 vector is available with single or tandem affinity tags at the N- or C- terminus to facilitate protein purification and detection. The pT7CFE vectors are suitable for insertion of cloned genes, cDNAs, ORFs or PCR products for in vitro transcription and translation.
  • Optimised performance designed to provide the highest yield in the human in vitro translation system
  • Many options multiple tag and tag-location options available
  • Modular MCS multiple cloning site is maintained across vector family to facilitate subcloning
  • Cleavable tags HRV 3C cleavage site available on select vectors
pT7CFE1 expression vectors contain the Encephalomyocarditis virus (EMCV) internal ribosome entry site (IRES) element that is critical for high levels of cap-independent protein expression in the Human In Vitro Translation System. Each vector features a highly-compatible multiple cloning site (MCS) to facilitate easy insertion of protein coding sequences into and between vectors. The pT7CFE1 Vector is available with single or tandem affinity tags at the N- or C- terminus to facilitate protein purification and detection. The pT7CFE Vectors are suitable for insertion of cloned genes, cDNAs, ORFs or PCR products for in vitro transcription and translation. Custom cloning services are also available.