Nonidet® P 40 Substitute (NP-40) 10% in aqueous solution

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85124
PIER85124EA 217 GBP
PIER85124
Nonidet® P 40 Substitute (NP-40) 10% in aqueous solution
Nonidet® P 40 Substitute (NP-40)
MDL Number: MFCD00132851
CAS Number: 9016-45-9
EINECS: 500-024-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR