UV transilluminator bulbs for myECL™ imager
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
62255
PIER62255EA 81.2 GBP
PIER62255
UV transilluminator bulbs for myECL™ imager
Photodocumentation Systems
These lamp bulbs with ultraviolet emission are used to replace the one of six pre-installed 306 nm UV bulbs in the myECL™ imager. All six lamps must be the same bulb type (365 or 306 nm).