Formaldehyde 16% (w/v) methanol-free, Pierce™

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

28906 28908
PIER28906EA 66.7 GBP
PIER28906 PIER28908
Formaldehyde 16% (w/v) methanol-free, Pierce™
Formaldehyde

Methanol-free formaldehyde is a highly reactive, cell-permeable agent that is used by researchers as a reversible crosslinking agent for proteins and nucleic acids within the cell or as a general cell-fixing agent for imaging-based applications.


  • Formaldehyde solution remains stable and free of contamination in an amber pre-scored ampoule
  • Package sizes geared for applications ranging from culture-targeted cell fixing to in-cell protein-based applications


1 ml ampoules are intended for protein-based in cell applications such as chromatin immunoprecipitation (ChIP) or protein interaction discovery.
10 ml ampoules are intended for bioimaging (high content screening/analysis) such as fixing cells on a culture plate before image analysis.

Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 3287
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR