NHS-activated agarose, Pierce™
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
26198 26196 26197 26199 26200
PIER26198EA 383 GBP
PIER26198 PIER26196 PIER26197 PIER26199 PIER26200
NHS-activated agarose, Pierce™
Protein Purification Protein Purification Resins
NHS-activated agarose is an amine-reactive, beaded-agarose resin for rapid and stable immobilisation of proteins, peptides and other ligands via primary amines.
  • Immobilise in a simple one-step reaction
  • No hazardous chemicals needed
  • Available as dry powder, spin columns or slurry
  • Leak-resistant chemistry makes it resuable
  • Coupling capacity >25 mg/ml for dry resin and >30 mg/ml for slurry
  • Affinity resin is adaptable to column and batch affinity chromatography techniques and FPLC applications
NHS-activated agarose is crosslinked, 6% beaded agarose resin that contains N-hydroxysuccinimide (NHS) functional groups. The activated resin reacts with primary amines to form stable amide linkages that covalently immobilise antibodies or other proteins for use in affinity purification procedures.