Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), Pierce™

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

24510 24525
PIER24510EA 451 GBP
PIER24510 PIER24525
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), Pierce™
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)

Pierce™ Sulfo-NHS (N-hydroxysulfosuccinimide) is a chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters for bioconjugation, crosslinking, labeling and immobilisation methods.


  • Improve efficiency of EDC coupling reactions
  • Water-soluble and membrane-impermable
  • Activate nanoparticles with amine-reactive Sulpho-NHS esters
  • Crosslink proteins to carboxyl-coated beads or surfaces more efficiently


Derivatives of Sulpho-NHS are easily synthesised by mixing the Sulpho-NHS with a carboxyl-containing molecule and a dehydrating agent. The method is the basis for generating many types of protein labeling reagents, including amine-reactive fluorescent dyes, biotin affinity tags and pegylation compounds.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₄H₄NNaO₆S
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now

Specification Test Results

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Carbodiimide
Spacer Short
Form Powder
Quantity 500 mg
Type Crosslinker
Solubility Water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR