Capillary columns, Elite-Guard and Elite-Siltek® for autosystem
Supplier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N9316608
PERCN9316608EA 187 GBP
PERCN9316608
Capillary columns, Elite-Guard and Elite-Siltek® for autosystem
Chromatography Columns
These guard columns lengthen the life of the capillary column and improve the analyte focusing.
  • Increase the life of the capillary column and performance
  • Maintains retention time and resolution on capillary column
  • Traps non-volatile particles
Both the intermediate-polarity deactivated guard columns and the Siltek® deactivated guard columns are important to match the polarity of the solvent and the polarity of the surface deactivation. Temperature range is 325 °C for Elite-Guardand 380 °C for Elite-Siltek® guard.