Capillary columns, Elite-Molesieve for autosystem
Supplier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N9316361
PERCN9316361EA 1850 GBP
PERCN9316361
Capillary columns, Elite-Molesieve for autosystem
Chromatography Columns
Elite-Molesieve PLOT phase capillary columns are suitable for the analysis of permanent gases.
Order Now
SPECIFICATIONS
Environmentally Preferable