Sulphur in polysulphide oil standard solutions
Supplier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N9308724 N9308714 N9308715 N9308716 N9308717 N9308718 N9308719 N9308720 N9308722 N9308723 N9308721 N9308394 N9308393 N9308392 N9308391 N9308390 N9308395
PERCN9308724EA 549 GBP
PERCN9308724 PERCN9308714 PERCN9308715 PERCN9308716 PERCN9308717 PERCN9308718 PERCN9308719 PERCN9308720 PERCN9308722 PERCN9308723 PERCN9308721 PERCN9308394 PERCN9308393 PERCN9308392 PERCN9308391 PERCN9308390 PERCN9308395
Sulphur in polysulphide oil standard solutions
Standards Cationic Standards Sulphur Standards
Polysulphide oil products offer high purity, low volatility and minimal diffusion through sample films.
These standards are well suited to ultra low level sulphur analysis, quality control, drift correction, blank measurement and calibration standards for XRF and other sulphur analysis techniques.