Aluminium headspace caps
Supplier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B0159356 N9303350
PERCB0159356EA 92.6 GBP
PERCB0159356 PERCN9303350
Aluminium headspace caps
Closures Chromatography Vial Closures
Un-Assembled aluminum caps and septa for all of your analytical and biological applications.
  • Temperature resistance up to 130 °C
  • Very economical and has low resistance to coring