Biopharm pressure sensors, SciLog®
Supplier: PARKER HANNIFIN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
080-699PSX-5
PARK080-699PSX-5EA 1230 GBP
PARK080-699PSX-5
Biopharm pressure sensors, SciLog®
Fluid Sensors
SciLog® range of pre-calibrated, single-use sensors, can measure and document pressure in a range of filtration, separation and purification applications.
  • Trans-membrane pressure (TMP) control
  • Suitable for single-use GMP applications
  • The sensors are validated for exposure to gamma irradiation, autoclave and sodium hydroxide sterilization
Its applications are growth media preparation, bioreactor maintenance, cell culture harvest, buffer preparation, product filtration, tangential flow filtration, virus control and bulk drug dispense.