Feeder cabinets
Supplier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FFCS-0540-SST FFCS-1040-SST
PALBFFCS-0540-SSTEA 7280 GBP
PALBFFCS-0540-SST PALBFFCS-1040-SST
Feeder cabinets
Furniture Storage Cabinets
The feeder cabinets support the gowning operation. Made of stainless steel 304 with electropolished finish.
  • Front access doors are used for reloading and for storage purposes
  • The bottom cabinet can be used for storage or disposal
  • Perspex dispenser slots ensure that garments/disposables can be extracted one at a time and do not spill out
Delivery information: Typically available from stock for direct shipment from the supplier. Delivered as shipped assembled or flat packed.