Polyvinyl alcohol, fully hydrolysed, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

P1763-250G P1763-1KG
P1763-250GEA 69.6 GBP
P1763-250G P1763-1KG
Polyvinyl alcohol, fully hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol

Prepared by alkaline hydrolysis.

Formula: [-CH₂CH(OH)-]n
Density: 1.080 g/cm³
Storage Temperature: Room Temperature
MDL Number: MFCD00081922
CAS Number: 9002-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21668879363dc296d25868f5b817236c9ee77516&vid=c9f3d32b556865c1a3e8ed6213306016b6768acb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) White to Off-White
Appearance (Form) Powder
Solubility (Colour) Colorless to Faint Yellow
Solubility (Turbidity)
50 mg/mL, H2O
Clear
Viscosity
4 % Aqueous solution at 20 deg C (Hoeppler Method)
11.6 - 15.4 cps
Hydrolysis 99.2 - 99.7 mole%
Recommended Retest Period 4 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR