(±)-1,2-Propanediol Ph. Eur., Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P148-33EA 61.4 GBP
P148-33 P148-38
(±)-1,2-Propanediol Ph. Eur., Macron Fine Chemicals™
Propylene glycol
Formula: CH₃CH(OH)CH₂OH
MDL Number: MFCD00064272
CAS Number: 57-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR