Urea Ph. Eur., Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P123-38EA 1550 GBP
P123-38
Urea Ph. Eur., Macron Fine Chemicals™
Urea
Formula: NH₂CONH₂
MDL Number: MFCD00008022
CAS Number: 57-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR