mRNA transfection reagents, RmesFect™
Supplier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
RM21000 RM20500
OZBIRM21000EA 355 GBP
OZBIRM21000 OZBIRM20500
mRNA transfection reagents, RmesFect™
Transfection Reagents
RmesFect™ transfection reagent is based on the TEE-technology and is specifically designed for mRNA transfection with high efficiency and low toxicity. It is suitable for in vitro mRNA (dsRNA) transfection in a large variety of cells.
  • Highly efficient, achieve mRNA delivery in all cells
  • Ready to use, no need for additional buffer
  • Low nucleic acid amount, minimised toxicity
  • Protects mRNA against degradation
  • Compatible with any culture medium
  • Serum compatible, medium change not required
mRNA transfection provides several advantages over plasmid DNA delivery, no need for nuclear uptake - mRNA translation into proteins occurs in the cytoplasm, faster protein expression than DNA transfection, no genomic integration, Protein expression in a total promoter-independent manner.
Store at –20 °C.
Delivery information: Shipped at room temperature.
Order Now
SPECIFICATIONS
Environmentally Preferable
mRNA Yes